Merck
  • Pharmacokinetic and pharmacodynamic analysis of inosine monophosphate dehydrogenase activity in hematopoietic cell transplantation recipients treated with mycophenolate mofetil.
Pharmacokinetic and pharmacodynamic analysis of inosine monophosphate dehydrogenase activity in hematopoietic cell transplantation recipients treated with mycophenolate mofetil.
Biology of blood and marrow transplantation : journal of the American Society for Blood and Marrow Transplantation (2014-04-15)
Hong Li, Donald E Mager, Brenda M Sandmaier, Barry E Storer, Michael J Boeckh, Meagan J Bemer, Brian R Phillips, Linda J Risler, Jeannine S McCune
ABSTRACT
A novel approach to personalizing postgrafting immunosuppression in hematopoietic cell transplantation (HCT) recipients is evaluating inosine monophosphate dehydrogenase (IMPDH) activity as a drug-specific biomarker of mycophenolic acid (MPA)-induced immunosuppression. This prospective study evaluated total MPA, unbound MPA, and total MPA glucuronide plasma concentrations and IMPDH activity in peripheral blood mononuclear cells (PMNCs) at 5 time points after the morning dose of oral mycophenolate mofetil (MMF) on day +21 in 56 nonmyeloablative HCT recipients. Substantial interpatient variability in pharmacokinetics and pharmacodynamics was observed and accurately characterized by the population pharmacokinetic-dynamic model. IMPDH activity decreased with increasing MPA plasma concentration, with maximum inhibition coinciding with maximum MPA concentration in most patients. The overall relationship between MPA concentration and IMPDH activity was described by a direct inhibitory maximum effect model with an IC50 of 3.23 mg/L total MPA and 57.3 ng/mL unbound MPA. The day +21 IMPDH area under the effect curve (AUEC) was associated with cytomegalovirus reactivation, nonrelapse mortality, and overall mortality. In conclusion, a pharmacokinetic-dynamic model was developed that relates plasma MPA concentrations with PMNC IMPDH activity after an MMF dose in HCT recipients. Future studies should validate this model and confirm that day +21 IMPDH AUEC is a predictive biomarker.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Metanolo, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanolo, ACS reagent, ≥99.8%
Sigma-Aldrich
Idrossido di ammonio, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Potassio cloruro, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Sodio idrossido, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodio idrossido, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Metanolo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Idrossido di sodio, 50% in H2O
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammonio acetato, ACS reagent, ≥97%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Idrossido di ammonio, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Metanolo, anhydrous, 99.8%
Sigma-Aldrich
Idrossido di sodio, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Ammonio acetato, ≥99.99% trace metals basis
Supelco
Ammonio acetato, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Potassio cloruro, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Idrossido di sodio, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Sodio idrossido, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Potassio cloruro, Molecular Biology, ≥99.0%
Sigma-Aldrich
Sodio idrossido, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Metanolo, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Potassio cloruro, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%