Merck
  • Comparison of capillary electrophoresis-mass spectrometry and hydrophilic interaction chromatography-mass spectrometry for anionic metabolic profiling of urine.
Comparison of capillary electrophoresis-mass spectrometry and hydrophilic interaction chromatography-mass spectrometry for anionic metabolic profiling of urine.
Talanta (2014-12-06)
Miranda G M Kok, Govert W Somsen, Gerhardus J de Jong
ABSTRACT
In order to assess the utility of a recently developed capillary electrophoresis-mass spectrometry (CE-MS) method for the study of anionic metabolites in urine, a comparison was made with hydrophilic interaction chromatography-MS (HILIC-MS) using negative electrospray ionization. After optimization of the HILIC conditions, a gradient employing 10mM ammonium acetate (pH 6.8) in acetonitrile-water (5 min 90% acetonitrile followed by 90%-50% acetonitrile in 10 min) was selected, providing baseline separation of five representative anionic test metabolites. Relative standard deviations (RSDs) for HILIC retention times and peak areas were below 0.2% and 7.7%, respectively, and detection limits were in the range 0.04-2.21 μM. Metabolites in rat urine could also be analysed in a reproducible way with retention time and peak area RSDs below 0.6% and 13.6%, respectively. The CE-MS and HILIC-MS methods were compared in terms of reproducibility, sensitivity, selectivity and coverage of the anionic urinary metabolome. In general, peak area RSDs were similar whereas HILIC-MS yielded better retention-time repeatability and up to 80 times lower detection limits (expressed in injected concentration) for test metabolites as compared to CE-MS. Rat urine analysis by HILIC-MS provided detection of 1360 molecular features compared to 347 molecular features revealed with CE-MS. Of these, a number of 144 molecular features were found with both HILIC-MS and CE-MS, which showed on average 10 times higher peak areas in HILIC-MS. The HILIC retention and CE migration times of the common features were clearly not correlated. The HILIC and CE behavior of the test metabolites and 16 putatively identified common features were evaluated involving their physicochemical properties, indicating a markedly different separation selectivity, and thus significant degree of orthogonality of HILIC and CE.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Metanolo, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanolo, ACS reagent, ≥99.8%
Sigma-Aldrich
Idrossido di ammonio, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Acido acetico, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Sodio idrossido, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Acido acetico, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acido formico, reagent grade, ≥95%
Sigma-Aldrich
Sodio idrossido, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Metanolo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Idrossido di sodio, 50% in H2O
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammonio acetato, ACS reagent, ≥97%
Sigma-Aldrich
Acido formico, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Idrossido di ammonio, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Acido formico, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Supelco
Formato di ammonio, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Trietilammina, ≥99.5%
Sigma-Aldrich
Metanolo, anhydrous, 99.8%
Sigma-Aldrich
Idrossido di sodio, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Formato di ammonio, reagent grade, 97%
Sigma-Aldrich
Ammonio acetato, ≥99.99% trace metals basis
Supelco
Ammonio acetato, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Idrossido di sodio, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acido acetico, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)