Changer de vue
| Taille/Référence | Disponibilité | Prix |
|---|
A propos de cet article
NACRES:
NB.43
UNSPSC Code:
41121800
eCl@ss:
32160703
Capacity:
1,000 mL
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aiderjoint
joint stopper27
manufacturer/tradename
Corning 3360-1LFO
capacity
1,000 mL
General description
A general purpose batch distillation assembly with the facility or addition via the Pyrex stoppered neck. Ideally suited for the batch production of high purity distilled water. The flask has a flat bottom with a side delivery tube, terminating in a joint. Condenser tubulations are 10mm O.D.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Still not finding the right product?
Explore all of our products under Pyrex® general purpose distilling apparatus, Pyrex stopper, ST joint