Changer de vue
| Taille/Référence | Disponibilité | Prix |
|---|---|---|
1 ea | Vérifier la disponibilité des articles du panier | 126,00 € |
A propos de cet article
NACRES:
NB.51
UNSPSC Code:
41121809
Joint:
joint: ST/NS top19/22
Packaging:
pkg of 1 ea
126,00 €
Vérifier la disponibilité des articles du panier
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aiderjoint
joint: ST/NS top19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F479250A
capacity
250 mL , 250 mL
General description
Supplied with a Glass or PTFE stopper
Legal Information
Synthware is a trademark of Kemtech America Inc.
1 of 1
Cet article | |||
|---|---|---|---|
| description 250 mL, top joint: ST/NS 19/22 | description 250 mL, top joint: ST/NS 19/22 | description 50 mL, top joint: ST/NS 19/22 | description 250 mL, Joints: (22) |
| manufacturer/tradename Synthware F479250A | manufacturer/tradename Synthware F479250 | manufacturer/tradename Synthware F479050 | manufacturer/tradename Synthware F472250A |
| capacity 250 mL, 250 mL | capacity 250 mL, 250 mL | capacity 50 mL , 50 mL | capacity 250 mL, 250 mL |
| joint joint: ST/NS top19/22 | joint joint: ST/NS top19/22 | joint joint: ST/NS top19/22 | joint Joints: (22) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ Squibb-style separatory funnel with PTFE stopcock and Glass stopper