Merck
  • Influence of protein and carbohydrate contents of soy protein hydrolysates on cell density and IgG production in animal cell cultures.
Influence of protein and carbohydrate contents of soy protein hydrolysates on cell density and IgG production in animal cell cultures.
Biotechnology progress (2015-06-18)
Abhishek J Gupta, Peter A Wierenga, Harry Gruppen, Jan-Willem Boots
ABSTRACT
The variety of compounds present in chemically defined media as well as media supplements makes it difficult to use a mechanistic approach to study the effect of supplement composition on culture functionality. Typical supplements, such as soy protein hydrolysates contain peptides, amino acids, carbohydrates, isoflavones, and saponins. To study the relative contribution of these compound classes, a set of hydrolysates were produced, containing 58-83% proteinaceous material and 5-21% carbohydrates. While the content of the different compounds classes varied, the composition (e.g., peptide profiles, carbohydrate composition) did not vary in hydrolysates. The hydrolysates were supplemented to a chemically defined medium in cell culture, based on equal weight and on equal protein levels. The latter showed that an increase in the carbohydrate concentration significantly (P value < 0.004) increased integral viable cell density (IVCD) (R = 0.7) and decreased total IgG (R = -0.7) and specific IgG production (R = -0.9). The extrapolation of effects of protein concentration showed that an increase in protein concentration increased total and specific IgG production and suppressed IVCD. In addition to proteins and carbohydrates, the functionality of soy protein hydrolysates may be modulated by the presence of other minor compounds. In the current study, the large differences in the balance between total proteins and total carbohydrates in the supplemented media seem to be a main factor influencing the balance between the viable cell density, total IgG, and specific IgG production.
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65 IU/mg
Sigma-Aldrich
TWEEN® 20, viscous liquid, suitable for cell culture
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Daidzein, ≥98%, synthetic