Merck
  • Lessons learned from flame retardant use and regulation could enhance future control of potentially hazardous chemicals.
Lessons learned from flame retardant use and regulation could enhance future control of potentially hazardous chemicals.
Health affairs (Project Hope) (2011-05-11)
Phil Brown, Alissa Cordner
ABSTRACT
Flame retardant chemicals, used in many furniture and electronic products to slow combustion, are an example of science and policy problems related to chemical usage. Drawing on a literature review and interviews with stakeholders, we argue that early warnings of the health hazards some flame retardants pose were not heeded as their use expanded to different products. Continued use of flame retardants coincides with their increased presence in people and adverse human health effects. We recommend policy changes applicable to oversight of chemicals in general: examining classes of chemicals rather than individual ones, assessing alternative methods and materials, product labeling, and stronger regulation.