Merck
Antioxidant vitamins and magnesium and the risk of hearing loss in the US general population.
The American journal of clinical nutrition (2013-11-08)
Yoon-Hyeong Choi, Josef M Miller, Katherine L Tucker, Howard Hu, Sung Kyun Park
ABSTRACT
The protective effects of antioxidant vitamins on hearing loss are well established in animal studies but in few human studies. Recent animal studies suggest that magnesium intake along with antioxidants may act in synergy to prevent hearing loss. We examined associations between intake of antioxidant vitamins (daily β-carotene and vitamins C and E) and magnesium and hearing thresholds and explored their joint effects in US adults. We analyzed cross-sectional data from 2592 participants aged 20-69 y from NHANES 2001-2004. Hearing thresholds as pure tone averages (PTAs) at speech (0.5, 1, 2, and 4 kHz) and high frequencies (3, 4, and 6 kHz) were computed. When examined individually, modeled as quartiles, and after adjustment for potential confounders, higher intakes of β-carotene, vitamin C, and magnesium were associated with lower (better) PTAs at both speech and high frequencies. High intakes of β-carotene or vitamin C combined with high magnesium compared with low intakes of both nutrients were significantly associated with lower (better) PTAs at high frequencies (-14.82%; 95% CI: -20.50% to -8.74% for β-carotene + magnesium and -10.72%; 95% CI: -16.57% to -4.45% for vitamin C + magnesium). The estimated joint effects were borderline significantly larger than the sums of the individual effects [high β-carotene/low magnesium (-4.98%) and low β-carotene/high magnesium (-0.80%), P-interaction = 0.08; high vitamin C/low magnesium (-1.33%) and low vitamin C/high magnesium (2.13%), P-interaction = 0.09]. Dietary intakes of antioxidants and magnesium are associated with lower risks of hearing loss.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acido L-ascorbico, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Acido L-ascorbico, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Acido L-ascorbico, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Acido L-ascorbico, 99%
Sigma-Aldrich
(+)- L-ascorbato di sodio, crystalline, ≥98%
Sigma-Aldrich
Acido L-ascorbico, reagent grade, crystalline
Supelco
Acido L-ascorbico, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(+)- L-ascorbato di sodio, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
(+)- L-ascorbato di sodio, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
β-Carotene, synthetic, ≥95% (HPLC), crystalline
Sigma-Aldrich
β-Carotene, synthetic, ≥93% (UV), powder
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Acido L-ascorbico, ACS reagent, ≥99%
Supelco
Acido L-ascorbico, analytical standard
Sigma-Aldrich
Acido L-ascorbico, meets USP testing specifications
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Acido L-ascorbico, reagent grade
Supelco
β-Carotene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
Acido L-ascorbico, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Acido L-ascorbico, FCC, FG
Supelco
(+)- L-ascorbato di sodio, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acido L-ascorbico, BioUltra, ≥99.5% (RT)
Acido L-ascorbico, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Acido L-ascorbico, puriss. p.a., ≥99.0% (RT)
Supelco
Acido L-ascorbico, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%