Merck
Bicalutamide treatment for locally advanced prostate cancer.
Hospital medicine (London, England : 1998) (2000-10-26)
J Anderson
ABSTRACT
In men with locally advanced prostate cancer, bicalutamide 150 mg monotherapy provides a similar disease outcome to medical or surgical castration. However, castration is associated with loss of sexual interest and function, decreased energy and an increased risk of osteoporotic fractures. Bicalutamide 150 mg monotherapy has less impact on sexual interest and physical capacity than castration.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Bicalutamide (CDX), ≥98% (HPLC), powder
Prezzi e disponibilità al momento non sono disponibili
Bicalutamide, European Pharmacopoeia (EP) Reference Standard
Prezzi e disponibilità al momento non sono disponibili
Bicalutamide for system suitability, European Pharmacopoeia (EP) Reference Standard
Prezzi e disponibilità al momento non sono disponibili