Merck
Long-acting isosorbide mononitrate.
Drugs (1999-02-10)
A Prakash, A Markham
ABSTRACT
Elantan long (EL) is the only long-acting isosorbide mononitrate formulation based on a controlled membrane principle. 30% of the dose is available for immediate release and the remaining 70% is gradually released over time. After oral administration, isosorbide mononitrate EL has a quick onset of action (30 minutes) and effects are evident for up to 17 hours. The antianginal effects of once-daily isosorbide mononitrate EL 50 to 100mg were generally similar to those of conventional isosorbide mononitrate or isosorbide dinitrate 10 to 20mg twice or 3 times daily, or sustained-release nifedipine 20mg twice or 3 times daily. Patients tended to show a better response with once-daily isosorbide mononitrate EL 50mg than with bupranolol 100mg. Patients showed significantly greater improvement in some quality-of-life indices with once-daily isosorbide mononitrate EL than with twice or 3 times daily regimens of conventional isosorbide mononitrate or isosorbide dinitrate. Tolerance did not develop after 13 months of once-daily administration of isosorbide mononitrate EL. No rebound increase in incidence of ischaemic episodes was observed after discontinuation of the drug.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Isosorbide mononitrate, European Pharmacopoeia (EP) Reference Standard
Prezzi e disponibilità al momento non sono disponibili