mAbs Upstream
Products for Development and Manufacturing.
The upstream workflow encompasses all steps leading to cell harvest including optimization of culture conditions, cell line and product characterization, media development, upstream process chemicals; and scale-up of the batch, fed-batch or perfusion process to larger bioreactors. The goal of upstream biomanufacturing is to increase cell density and product titer.
Sterile Filtration
A high risk of contamination exists in upstream processing due to nutrient-rich environments. In the upstream workflow, bioburden control focuses on sourcing and characterization of raw materials to prevent introduction of adventitious agents into cell culture operations. In addition, cell culture media is sterilized, often sterile-filtered, or processed through filters that remove bacteria and mycoplasma, to prevent them from entering the bioreactor. Prefiltration extends the life of sterilizing filter by removing particulates that would result in early plugging of the sterilizing filter.
Our industry-leading portfolio of sterilizing filters includes:
- Millipore® Filters (buffer and media filtration, sterile filtration)
- Millipore® Equipment and Systems (housings)
- Millipore® Services (single-use and filter validation, system services, equipment installation, system qualification, system training)
- Millipore® Automation and Analytics Software
Related Products
Response not successful: Received status code 500
Related Categories
A cell line that maintains favorable growth and productivity is vital for mab manufacturing.
Our cell culture media makes your intensified or fed-batch processes more robust and efficient.
Our recombinant, non-animal origin (NAO) supplements are formulated for cell culture applications.
Mobius® single-use mixing solutions include a complete range of mixers from 10 L to 3,000 L.
The Mobius® Bioreactor family accommodates batch, fed-batch, and perfusion upstream processes.
Our broad portfolio of membrane filters offers options for efficient processing of every pharma and biopharma stream.
Related Resources
- Brochure: Mission Control: Strategies for Effective Bioburden and Aseptic Control
In this series of articles, we provide information to highlight the differing objectives for bioburden control by unit operation and discuss the parameters that guide filter selection and optimization.
- Tech Note: Holistic Buffer Preparation
Buffer preparation is critical in biologics production, requiring significant volume and precise timing. The document will discuss optimizing in-house preparation and potential outsourcing, considering various options based on process needs.
- Application Note: Cell Culture Media Mixing and Microfiltration: Reducing Risk with Single-use Technology
This application note provides an overview of considerations for mixing reconstituted powdered cell culture media and filtration of the reconstituted media using sterilizing or mycoplasma removal filters.
- Tech Note: How Membrane Prefilters Enhance BioProcessing Efficiency
Milligard® PES filters with two layers of asymmetric polyethersulfone (PES) membranes are widely used in biopharmaceutical production to remove particulates from process fluids. When used as prefilters they can increase the capacity of sterilizing filters and improve process economics.
- Application Note: Cell Culture Media Filtration – Filter Selection and Sizing
This document discusses the trend in bioprocessing to enhance productivity upstream through improved cell culture media processing, highlighting the challenges posed by concentrated and complex media that can lead to premature fouling of membrane filters. It emphasizes the importance of selecting the right membrane filter, particularly addressing microbial retention, and explores the trade-offs between different filter options such as traditional sterilizing-grade filters and mycoplasma-retentive filters in minimizing bioreactor contamination risk.
- Technical Note: Demonstrating Millipore Express® SHR Mycoplasma Clearance Capability at Pilot and Production Scale over an Extended Filtration
Two studies were conducted to assess the mycoplasma retentive properties of Millipore Express® SHR with Prefilter filters using Acholeplasma laidlawii ATCC® 23206™, simulating typical processing parameters and operating pressures. The studies demonstrated that the filters, in both production scale and pilot scale configurations, were fully retentive when challenged with A. laidlawii at low organism challenge levels over extended filtration times.
- Technical Note: Mycoplasma Clearance by Millipore Express® SHR Filters at Pilot and Production Scales
The Millipore Express® SHR filters, equipped with a 0.1 µm polyethersulfone (PES) sterilizing membrane, offer enhanced microbial risk reduction, particularly targeting mycoplasma contaminants in cell culture media. Retention studies demonstrated that both pilot and production-scale filters consistently provided the expected level of mycoplasma retention over extended filtration times, affirming their performance under continuous flow processing conditions.
- Tech Note: Integrity Test Troubleshooting - Beyond Rewet and Retest
Integrity testing is crucial for sterilizing grade filters in biopharmaceutical processing. This application note aims to ensure accurate integrity testing and address specific challenges.
- Application Note: Cell Culture Media Filtration - Evaluating Cell Culture Performance
This document addresses the challenge of efficiently processing highly concentrated and complex cell culture media, emphasizing the benefits of membrane filtration in reducing the risk of bioreactor contamination. It discusses key considerations in selecting membrane filters based on microbial retention, compatibility, and capacity for specific media.
Explore Our Solutions