現在、価格および在庫状況を閲覧できません。 製品に関するお問い合わせはこちら(テクニカルサービス)
Inchi Code
1S/C6H14N2.2ClH/c1-3-8(2)6-4-7-5-6;;/h6-7H,3-5H2,1-2H3;2*1H
Inchi Key
CFHADJUBHYSVOF-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under N-Ethyl-N-methylazetidin-3-amine dihydrochloride