grade
certified reference material
agency
NMI Australia certified reference material
form
solid
manufacturer/tradename
NMI Australia
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
neat
storage temp.
2-8°C
InChI
1S/C20H10Cl2F5N3O3/c21-11-5-4-10(29-19(32)30-17(31)16-13(23)2-1-3-14(16)24)7-15(11)33-18-12(22)6-9(8-28-18)20(25,26)27/h1-8H,(H2,29,30,31,32)
InChI key
YOWNVPAUWYHLQX-UHFFFAOYSA-N
注意喚起語
Warning
ハザードコード
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
保管分類
13 - Non Combustible Solids
WGK
WGK 2
引火点 (F)
Not applicable
引火点(°C)
Not applicable