コンテンツへスキップ
Merck
  • Search for skin-whitening agent from Prunus plants and the molecular targets in melanogenesis pathway of active compounds.
Search for skin-whitening agent from Prunus plants and the molecular targets in melanogenesis pathway of active compounds.
Natural product communications (2014-04-03)
Kazuya Murata, Keisuke Takahashi, Haruka Nakamura, Kimihisa Itoh, Hideaki Matsuda
要旨
In our research program for discovering novel skin-whitening materials, screening of extracts from flowers of some Prunus species was performed using an anti-tyrosinase assay. Among the tested plants, the flowers of P. persica showed the most potent inhibitory activity. In addition, P. persica also showed suppression of melanogenesis in B16 rat melanoma cells. The active principles of tyrosinase inhibition and suppression of melanogenesis were revealed to be an afzelin (3-O-alpha-L-rhamnosylkaempferol) and a flavanone, naringenin. The mechanism of the anti-melanogenesis effect of these two compounds was disclosed, for the first time, as the suppression of the expression of tyrosinase protein, which was controlled by the inhibition of phosphorylation of p38 MAPK. These findings show that these compounds could be candidates for the novel molecular target for a skin-whitening agent.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
チロシナーゼ from mushroom, lyophilized powder, ≥1000 unit/mg solid
現在、価格および在庫状況を閲覧できません。