コンテンツへスキップ
Merck
  • Enhanced Antitumor Efficacy and Reduced Toxicity of Docetaxel Loaded Estradiol Functionalized Stealth Polymeric Nanoparticles.
Enhanced Antitumor Efficacy and Reduced Toxicity of Docetaxel Loaded Estradiol Functionalized Stealth Polymeric Nanoparticles.
Molecular pharmaceutics (2015-09-17)
Sanyog Jain, Gollapalli Spandana, Ashish Kumar Agrawal, Varun Kushwah, Kaushik Thanki
要旨
In spite of extensive research over the decades, breast cancer treatment is still a major challenge due to nonspecific distribution of the chemotherapeutics. This void can be filled by restricting the distribution of chemotherapeutics toward the cancerous cells. In the present report estradiol (E2) functionalization of docetaxel (DTX) loaded PLGA nanoparticles was supposed to have specific distribution of DTX to cancerous cells simultaneously avoiding the nonspecific distribution toward the normal cells. In line, E2-PEG-PLGA conjugate was synthesized and characterized by FTIR and NMR spectroscopy. Extensive optimization of different process variables resulted in the formation of spherical E2-PEG-PLGA NPs in the size range of 228.5 ± 11.8 nm and entrapment efficiency of 94.25 ± 2.49. Trehalose (5% w/v) resulted in the formation of a fluffy, easy to redisperse freeze-dried cake of nanoparticles. PXRD analysis revealed the amorphous nature of DTX encapsulated within the nanoparticles. X-ray photoelectron spectroscopy confirmed the presence of E2 over the surface of nanoparticles. In line with our hypothesis, cellular uptake studies on ER positive MCF-7 cells revealed relatively higher uptake and efficient localization into the nuclear region of E2-PEG-PLGA NPs in comparison with plain counterparts, while in the case of ER negative HeLa cells E2-PEG-PLGA NPs showed no difference in fluorescence pattern as compared to MCF-7 cells incubated with unmodified nanoformulation, indicating nonspecific delivery of DTX. Moreover, MTT assay revealed relatively higher cytotoxicity of E2-PEG-PLGA NPs in comparison with free DTX. Furthermore, in vivo pharmacokinetic studies revealed 9.36- and 4.79-fold enhancement in circulation half-life and AUC(0-∞), respectively, of E2-PEG-PLGA NPs in comparison with Taxotere. In vivo antitumor efficacy in DMBA induced rat model demonstrated significant reduction in tumor volume in comparison with the plain counterpart (PLGA-NPs) and a marketed formulation, Taxotere. Moreover, the safety of the estradiol functionalized PLGA NPs was confirmed by hepato- and nephrotoxicity studies.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
塩酸, ACS reagent, 37%
Sigma-Aldrich
ジエチルエーテル, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
ジクロロメタン, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
リン酸ナトリウム 二塩基性, ACS reagent, ≥99.0%
Sigma-Aldrich
重炭酸ナトリウム, ACS reagent, ≥99.7%
Sigma-Aldrich
ピリジン, ACS reagent, ≥99.0%
Sigma-Aldrich
塩酸, ACS reagent, 37%
Sigma-Aldrich
酢酸ナトリウム, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
塩化水素 溶液, 4.0 M in dioxane
Sigma-Aldrich
ジクロロメタン, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
ジエチルエーテル, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
重炭酸ナトリウム, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
重炭酸ナトリウム, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
酢酸ナトリウム, ACS reagent, ≥99.0%
Sigma-Aldrich
リン酸ナトリウム 二塩基性, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
ベンゼン, ACS reagent, ≥99.0%
Sigma-Aldrich
N-ヒドロキシスクシンイミド, 98%
Sigma-Aldrich
塩酸 溶液, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
チアゾリルブルーテトラゾリウムブロミド, 98%
Sigma-Aldrich
ピリジン, anhydrous, 99.8%
Sigma-Aldrich
チアゾリルブルーテトラゾリウムブロミド, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
リン酸ナトリウム 二塩基性, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
塩酸, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
塩酸, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
ジクロロメタン, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
塩化水素 溶液, 2.0 M in diethyl ether
Sigma-Aldrich
ピリジン, ReagentPlus®, ≥99%
Sigma-Aldrich
ジエチルエーテル, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
エチレンジアミン四酢酸, ACS reagent, 99.4-100.6%, powder