Merck
Efflux-mediated bis-indole resistance in Staphylococcus aureus reveals differential substrate specificities for MepA and MepR.
Bioorganic & medicinal chemistry (2010-03-02)
Timothy J Opperman, John D Williams, Chad Houseweart, Rekha G Panchal, Sina Bavari, Norton P Peet, Donald T Moir, Terry L Bowlin
ABSTRAKT
The bis-indoles are a novel class of compounds with potent antibacterial activity against a broad spectrum of Gram-positive and Gram-negative pathogens. The mechanism of action of these compounds has not been clearly defined. To study the mechanism of action of bis-indoles, selections for mutants of Staphylococcus aureus NCTC 8325 with reduced susceptibility to several chemically related bis-indoles were carried out using serial passages in subinhibitory compound concentrations. Resistant mutants were only obtained for one of the four bis-indoles tested (MBX-1090), and these appeared at concentrations up to 16X MIC within 10-12 passages. MBX-1090 resistance mutations produced a truncated open reading frame of mepR (SAOUHSC_00314), a gene encoding a MarR-like repressor. MepR regulates expression of mepA (SAOUHSC_00315), which encodes a member of the Multidrug and Toxic Compound Extrusion (MATE) family of efflux pumps. MBX-1090 resistance was reverted when mepR (wild type) was provided in trans. Microarray experiments and RT-PCR experiments confirmed that over-expression of mepA is required for resistance. Interestingly, MBX-1090 resistant mutants and strains overexpressing mepA from an expression vector did not exhibit cross-resistance to closely related bis-indole compounds. MBX-1090 did not induce expression of mepA, suggesting that this compound does not directly interact with MepR. Conversely, the bis-indoles that were not substrates of MepA strongly induced mepA expression. The results of this study suggest that MepA and MepR exhibit remarkably distinct substrate specificity for closely related bis-indoles.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Carbenicillin, Ready Made Solution, 100 mg/mL in ethanol/water
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Enrofloxacin, ≥99.0%
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Nalidixic acid, ≥98%
Supelco
Enrofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Supelco
Trimethoprim, VETRANAL®, analytical standard