Merck
Synthesis and biological evaluation of 3,7-diazabicyclo[4.3.0]nonan-8-ones as potential nootropic and analgesic drugs.
Journal of medicinal chemistry (2011-03-09)
Elisabetta Martini, Lorenzo Di Cesare Mannelli, Gianluca Bartolucci, Carlo Bertucci, Silvia Dei, Carla Ghelardini, Luca Guandalini, Dina Manetti, Serena Scapecchi, Elisabetta Teodori, Maria Novella Romanelli
ABSTRAKT
A series of cis and trans 3,7-diazabicyclo[4.3.0]nonan-8-ones has been synthesized and tested for their ability to revert scopolamine-induced amnesia in the mouse passive-avoidance test. The racemates of the most potent compounds 4 and 7 were separated and tested, but no enantioselectivity was found for the nootropic activity. Compounds 4 and 7 and their enantiomers displayed interesting antihyperalgesic activity in two models of neuropathic pain (streptozotocin-induced and oxalilplatin-induced neuropathy) in comparison with pregabalin.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Piracetam
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
Kwas(R)-(+)-α-liponowy, analytical standard