Merck
Risk of cancer among workers exposed to trichloroethylene: analysis of three Nordic cohort studies.
Journal of the National Cancer Institute (2013-06-01)
Johnni Hansen, Markku Sallmén, Anders I Seldén, Ahti Anttila, Eero Pukkala, Kjell Andersson, Ing-Liss Bryngelsson, Ole Raaschou-Nielsen, Jørgen H Olsen, Joseph K McLaughlin
ABSTRAKT
Trichloroethylene (TCE) is a widely used chlorinated solvent with demonstrated carcinogenicity in animal assays. Some epidemiologic studies have reported increased risk of cancer of the kidney, cervix, liver and biliary passages, non-Hodgkin lymphoma, and esophageal adenocarcinoma. We established a pooled cohort, including 5553 workers with individual documented exposure to TCE in Finland, Sweden, and Denmark. Study participants were monitored for the urinary TCE metabolite trichloroacetic acid from 1947 to 1989 and followed for cancer. Standardized incidence ratios (SIRs) were calculated based on cancer incidence rates in the three national populations. Cox proportionate hazard analyses were used for internal comparisons. Tests of statistical significance are two-sided. Overall, 997 cases of cancer (n = 683 in men; n = 314 in women) were identified during 154 778 person-years of follow-up. We observed statistically significant elevated standardized incidence ratios for primary liver cancer (1.93; 95% confidence interval [CI] = 1.19 to 2.95) and cervical cancer (2.31; 95% CI = 1.32 to 3.75). The standardized incidence ratio for kidney cancer was 1.01 (95% CI = 0.70 to 1.42) based on 32 cases; we did not observe a statistically significant increased risk of non-Hodgkin's lymphoma (SIR = 1.26; 95% CI = 0.89 to 1.73) or esophageal adenocarcinoma (SIR = 1.84; 95% CI = 0.65 to 4.65). Tobacco- and alcohol-associated cancers were not statistically significantly increased. Our results suggest TCE exposure is possibly associated with an increased risk for liver cancer. The relationship between TCE exposure and risks of cancers of low incidence and those with confounding by lifestyle and other factors not known in our cohort require further study.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Trichloroethylene solution, certified reference material, 5000 μg/mL in methanol
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Trichloroethylene, ACS reagent, ≥99.5%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Trichloroethylene, anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Trichloroethylene, analytical standard, stabilized with 30 – 50 ppm Diisopropylamine
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Informacje o cenach i dostępności nie są obecnie dostępne.