Merck
  • Effect of different concentrations of acetic, citric, and propionic acid dipping solutions on bacterial contamination of raw chicken skin.
Effect of different concentrations of acetic, citric, and propionic acid dipping solutions on bacterial contamination of raw chicken skin.
Poultry science (2013-07-23)
A Menconi, S Shivaramaiah, G R Huff, O Prado, J E Morales, N R Pumford, M Morgan, A Wolfenden, L R Bielke, B M Hargis, G Tellez
ABSTRAKT
Bacterial contamination of raw, processed poultry may include spoilage bacteria and foodborne pathogens. We evaluated different combinations of organic acid (OA) wash solutions for their ability to reduce bacterial contamination of raw chicken skin and to inhibit growth of spoilage bacteria and pathogens on skin during refrigerated storage. In experiment 1, raw chicken skin samples were dipped into a suspension of either 10(8) cfu/mL of Salmonella Typhimurium, Escherichia coli O157:H7, or Listeria monocytogenes for 30 s and then immersed in PBS or an OA wash solution mixture of 0.8% citric, 0.8% acetic, and 0.8% propionic acid (at equal wt/vol concentrations) for an additional 30 s. In experiment 2, three different concentrations of the OA wash solution (0.2, 0.4, and 0.6% at equal wt/vol concentrations) were tested against chicken skin samples contaminated with Salmonella Typhimurium. Viable pathogenic bacteria on each skin sample were enumerated after 1 and 24 h of storage at 4°C in both experiments. In experiment 3, skin samples were initially treated on d 1 with PBS or 2 concentrations of the OA mixture (0.4 and 0.8%), and total aerobic bacteria were enumerated during a 2-wk storage period. In all experiments, significant (P < 0.05) differences were observed when skin samples were treated with the OA wash solution and no spoilage organisms were recovered at any given time point, whereas increasing log10 numbers of spoilage organisms were recovered over time in PBS-treated skin samples. These results suggest that 0.2 to 0.8% concentrations of an equal-percentage mixture of this OA combination may reduce pathogens and spoilage organisms and improve food safety properties of raw poultry.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Propionic acid, ≥99.5%
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Supelco
Propionic acid, analytical standard
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~98%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG