Merck
Inhibition of the proton-coupled folate transporter (PCFT-SLC46A1) by bicarbonate and other anions.
Molecular pharmacology (2013-04-24)
Rongbao Zhao, Michele Visentin, Sylvia O Suadicani, I David Goldman
ABSTRAKT
The proton-coupled folate transporter (PCFT) plays a key role in intestinal folate absorption, and loss-of-function mutations in the gene encoding this transporter are the molecular basis for hereditary folate malabsorption. Using a stable transfectant with high expression of PCFT, physiologic levels of bicarbonate produced potent and rapidly reversible inhibition of PCFT-mediated transport at neutral pH. Bisulfite and nitrite also inhibited PCFT function at neutral pH, whereas sulfate, nitrate, and phosphate had no impact at all. At weakly acidic pH (6.5), bisulfite and nitrite exhibited much stronger inhibition of PCFT-mediated transport, whereas sulfate and nitrate remained noninhibitory. Inhibition by bisulfite and nitrite at pH 6.5 was associated with a marked decrease in the influx Vmax and collapse of the transmembrane proton gradient attributed to the diffusion of the protonated forms into these cells. Monocarboxylates such as pyruvate and acetate also collapsed the pH gradient and were also inhibitory, whereas citrate and glycine neither altered the proton gradient nor inhibited PCFT-mediated transport. These observations add another dimension to the unfavorable pH environment for PCFT function in systemic tissues: the presence of high concentrations of bicarbonate.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Pyruvic acid, 95%
Sigma-Aldrich
Pyruvic acid, 98%
Sigma-Aldrich
Pyruvic acid, natural, ≥95%, FG
Sigma-Aldrich
Pyruvic acid, ≥97%, FG
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, 99%
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%