Merck
  • Association of low baseline levels of erythrocyte folate with treatment nonresponse at three months in rheumatoid arthritis patients receiving methotrexate.
Association of low baseline levels of erythrocyte folate with treatment nonresponse at three months in rheumatoid arthritis patients receiving methotrexate.
Arthritis and rheumatism (2013-10-30)
M C F J de Rotte, P H P de Jong, S M F Pluijm, M Bulatović Calasan, P J Barendregt, D van Zeben, P A van der Lubbe, P B de Sonnaville, J Lindemans, J M W Hazes, R de Jonge
ABSTRAKT
To investigate whether baseline concentrations of one-carbon metabolism biomarkers are associated with treatment nonresponse and adverse events in rheumatoid arthritis (RA) patients receiving methotrexate (MTX). A prospective derivation cohort (n = 285) and validation cohort (n = 102) of RA patients receiving MTX were studied. Concentrations of plasma homocysteine, serum vitamin B12 , serum folate, erythrocyte vitamin B6 , and erythrocyte folate were determined at baseline and after 3 months of treatment. Nonresponse after 3 months was assessed using the Disease Activity Score in 28 joints (DAS28) and the European League Against Rheumatism (EULAR) response criteria. Adverse events at 3 months were assessed using biochemical parameters and health status questionnaires. Analyses were corrected for baseline DAS28, age, sex, MTX dose, comedications, and presence of the methylenetetrahydrofolate reductase 677TT genotype. In the derivation cohort, the mean DAS28 scores at baseline and 3 months were 4.94 and 3.12, respectively, and 78% of patients experienced adverse events. This was similar between the 2 cohorts, despite a lower MTX dose in the validation cohort. Patients with lower levels of erythrocyte folate at baseline had a higher DAS28 at 3 months in both the derivation cohort (β = -0.15, P = 0.037) and the validation cohort (β = -0.20, P = 0.048). In line with these results, lower baseline erythrocyte folate levels were linearly associated with a 3-month DAS28 of >3.2 in both cohorts (derivation cohort, P = 0.049; validation cohort, P = 0.021) and with nonresponse according to the EULAR criteria (derivation cohort, P = 0.066; validation cohort, P = 0.027). None of the other biomarkers (levels at baseline or changes over 3 months) were associated with the DAS28 or treatment nonresponse. Baseline levels of the biomarkers and changes in levels after 3 months were not associated with incidence of adverse events. A low baseline concentration of erythrocyte folate is associated with high disease activity and nonresponse at 3 months after the start of MTX treatment and could be used in prediction models for MTX outcome. None of the investigated one-carbon metabolism biomarkers were associated with incidence of adverse events at 3 months.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Homocysteine, ≥98.0% (NT)
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
Folic acid, meets USP testing specifications
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Supelco
Cyanocobalamin (B12), analytical standard
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Cyanocobalamin (Vitamin B12) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Methotrexate, European Pharmacopoeia (EP) Reference Standard
Folic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methotrexate solution, 1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®