Merck
  • Evaluation of the antioxidant effect of ganghwayakssuk (Artemisia princeps Pamp.) extract alone and in combination with ascorbic acid in raw chicken patties.
Evaluation of the antioxidant effect of ganghwayakssuk (Artemisia princeps Pamp.) extract alone and in combination with ascorbic acid in raw chicken patties.
Poultry science (2013-11-16)
K E Hwang, H W Kim, Y S Choi, S Y Lee, E J Yeo, Y K Ham, S M Choi, M A Lee, C J Kim
ABSTRAKT
We investigated the inhibition of lipid oxidation of raw chicken patties by the antioxidants ascorbic acid (Aa), ganghwayakssuk extracts (GE), and their combination (Aa + GE). All antioxidant combinations were effective at delaying lipid oxidation compared with the control or Aa. A combination of Aa + GE (0.05% Aa + 0.2% GE) was the most effective for delaying lipid oxidation (TBA reactive substances, conjugated dienes, and peroxide formation). The color values of all samples were significantly affected by adding GE. Additionally, the redness, color difference, and hue values of all treatments, except for Aa, were lower than those of the control as the amount of GE increased. The total viable bacterial counts of samples with GE 0.2 and Aa + GE 0.2 were significantly affected during storage (P < 0.05). The results suggest that adding an antioxidant combination reduced the oxidative stress and microbial growth of raw chicken patties stored for 12 d under normal refrigeration temperature, which may extend the shelf life of chicken patties.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard