Merck
  • Metabolic and histopathological profile of Rattus norvegicus (Wistar) experimentally infected by Angiostrongylus cantonensis (Chen, 1935).
Metabolic and histopathological profile of Rattus norvegicus (Wistar) experimentally infected by Angiostrongylus cantonensis (Chen, 1935).
Experimental parasitology (2013-12-18)
Juberlan Silva Garcia, Camila dos Santos Lúcio, Tatiane Cristina dos Santos Bonfim, Arnaldo Maldonado Junior, Victor Menezes Tunholi, Vinícius Menezes Tunholi-Alves, Esther Maria Mota, Raquel de Oliveira Simões, André Campos Santana, Cleber Hooper, Jairo Pinheiro, Marcio Neves Bóia
ABSTRAKT
Eosinophilic meningitis is a disease characterized by increased eosinophils in the cerebrospinal fluid (CSF), which is the most commonly caused by invasion of the central nervous system by helminths, as occurs in Angiostrongylus cantonensis infections. The rodent Rattus norvegicus is the definitive natural host and humans act as accidental hosts and can become infected by eating raw or undercooked snails or food contaminated with infective L3 larvae. Recently in Brazil there have been four cases of eosinophilic meningitis due to ingestion of infected Achatina fulica. To evaluate biochemical and histopathological changes caused by this parasite, R. norvegicus were experimentally infected with 100 L3 larvae of A. cantonensis. After the anesthetic procedure, serum from the rodents was collected from the inferior vena cava for evaluation of the levels of aspartate aminotransferase (AST), alanine aminotransferase (ALT), alkaline phosphatase (ALKP), gamma-glutamyl transferase (GGT), total protein and its fractions. During the necropsy, the liver was collected and weighed. Then a 1-g fragment was extracted from the major lobe to quantify the hepatic glycogen and fragment remainder was taken from the same lobe and fixed in Milloning's formalin for histopathological examination. Additionally, helminths were collected from the brain and lungs of the rodents. The activities of AST, ALT, ALKP and GGT in the serum and hepatic glycogen increased in response to infection, while the levels of globulin and total protein increased only in the eighth week of infection and there was a reduction in the levels of serum glucose. Albumin and bilirubin concentrations remained stable during the experiment. Infection with A. cantonensis caused metabolic and histopathological changes in the rodents. This study can contribute to a better understanding of the relationship between A. cantonensis and R. norvegicus.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
γ-Glutamyl Transferase, ERM®, certified reference material
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
γ-Glutamyltranspeptidase from equine kidney, Type VI, 5-12 units/mg solid
Sigma-Aldrich
γ-Glutamyltranspeptidase from porcine kidney, Type IV, 2.0-6.0 units/mg solid