Merck
Effect of temperature, pH and heme ligands on the reduction of Cygb(Fe(3+)) by ascorbate.
Archives of biochemistry and biophysics (2014-05-02)
Jianjing Tong, Joseph R Zweier, Rachael L Huskey, Raed S Ismail, Craig Hemann, Jay L Zweier, Xiaoping Liu
ABSTRAKT
Cytoglobin (Cygb) plays a role in regulating vasodilation in response to changes in local oxygen concentration by altering the rate of nitric oxide (NO) metabolism. Because the reduction of Cygb(Fe(3+)) by a reductant is the control step for Cygb-mediated NO metabolism, we examined the effects of temperature, pH, and heme ligands on the Cygb(Fe(3+)) reduction by ascorbate (Asc) under anaerobic conditions. The standard enthalpy of Cygb(Fe(3+)) reduction by Asc was determined to be 42.4 ± 3.1 kJ/mol. The rate of Cygb(Fe(3+)) reduction increased ~6% per °C when temperature varied from 35°C to 40°C. The yield and the rate of Cygb(Fe(3+)) reduction significantly increases with pH (2-3 times per pH unit), paralleling the formation of the Asc ion (A(2-)) and the increased stability of reduced state of heme iron at high pH values. Heme ligand cyanide (CN(-)) decreased the yield and the rate of Cygb(Fe(3+)) reduction, but ligands CO and NO allowed the process of Cygb(Fe(3+)) reduction to continue to completion. Critical information is provided for modeling and prediction of the process of Cygb-mediated NO metabolism in vessels in a range of temperature and pH values.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sodium ascorbate, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Kwas askorbinowy, United States Pharmacopeia (USP) Reference Standard