Merck
Class A scavenger receptor deficiency augments angiotensin II-induced vascular remodeling.
Biochemical pharmacology (2014-05-31)
Lingling Qian, Xiaoyu Li, Ru Fang, Zhuoyun Wang, Yiming Xu, Hanwen Zhang, Hui Bai, Qing Yang, Xudong Zhu, Jingjing Ben, Yong Xu, Qi Chen
ABSTRAKT
Class A scavenger receptor (SR-A) is a multifunctional molecule that participates in macrophage-mediated inflammation. Here we evaluated the role of SR-A in angiotensin II (Ang II)-induced hypertensive vascular remodeling. Chronic infusion of Ang II leads to an increased systolic blood pressure both in SR-A knockout (SR-A(-/-)) and wild type (SR-A(+/+)) mice with no significant difference between these two groups. SR-A(-/-) hypertensive mice, however, exhibited a marked augmentation of arterial wall thickening and vascular cell proliferation compared with SR-A(+/+) hypertensive mice. M1 macrophage markers were increased whereas M2 macrophage markers were decreased in vascular tissues of SR-A(-/-) mice. Co-culture experiments revealed that more pro-inflammatory cytokines like TNF-α were produced by SR-A(-/-) peritoneal macrophages leading to a stronger proliferation of primary vascular smooth muscle cells in vitro. In addition, SR-A(-/-) macrophages were more prone to lipopolysaccharide-induced M1 differentiation while resisting interleukin-4-induced M2 differentiation. Importantly, transplantation of SR-A(-/-) bone marrow into SR-A(+/+) mice significantly augmented Ang II-induced vascular remodeling. These results show that SR-A is critical for Ang II-induced vascular remodeling by regulating macrophage polarization. Therefore, SR-A may be a useful therapeutic target for the intervention of hypertensive vascular remodeling.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Angiotensin II human, ≥93% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Supelco
Acrylamide solution, 40% in H2O