Merck
  • Impact of thermal and organic acid treatment of feed on apparent ileal mineral absorption, tibial and liver mineral concentration, and tibia quality in broilers.
Impact of thermal and organic acid treatment of feed on apparent ileal mineral absorption, tibial and liver mineral concentration, and tibia quality in broilers.
Poultry science (2014-05-28)
A Hafeez, A Mader, F Goodarzi Boroojeni, I Ruhnke, I Röhe, K Männer, J Zentek
ABSTRAKT
Minerals play an important role for growth and bone stability in broilers. Thermal treatment and inclusion of organic acids in feed may affect the mineral absorption and tibial quality in broilers. The study was conducted to investigate the effect of thermal processing of feed including pelleting (P), long-term conditioning at 85°C (L), and expanding at 130°C (E) without and with 1.5% of an acid mixture containing 64% formic and 25% propionic acid on the apparent ileal absorption (AIA) of calcium, phosphorus, magnesium, potassium, sodium, iron, copper, manganese, and zinc, their concentrations in liver and tibia, as well as various tibial quality parameters in broilers. In total, 480 one-day-old Cobb broiler chicks were assigned using a completely randomized design with a 3 × 2 factorial arrangement. The ileal digesta, liver, and tibia were collected at d 35. The AIA of calcium and sodium was improved in group E compared with L (P ≤ 0.02 and P ≤ 0.01). Group P and E showed higher AIA for potassium than L (P ≤ 0.01). Bone ash content was increased in group E compared with L (P ≤ 0.04). The BW to bone weight ratio was lower and tibial zinc content was higher in group P compared with E (P ≤ 0.05). Tibial iron content was higher in group L than E (P ≤ 0.03). Acid addition did not affect AIA, mineral content in tibia, or tibial quality parameters. Thermal and acid treatment did not affect mineral concentrations in the liver, except an inconsistent interaction effect for DM content and sodium (P ≤ 0.03 and P ≤ 0.04, respectively). In conclusion, long-term thermal treatment reduced AIA of some minerals compared with short-term thermal treatments, but had no impact on tibia composition. Acid inclusion had no effect on AIA of minerals and tibia quality. Thermal treatment and the use of organic acids can therefore be considered as safe with regard to their impact on bone development in broilers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Thallium(I) formate, 97%
Supelco
Calcium formate, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Cesium formate, 98%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Calcium formate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium formate, 99.998% trace metals basis
Sigma-Aldrich
Sodium formate-13C, 99 atom % 13C
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Potassium formate, ReagentPlus®, 99%
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Potassium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Sodium formate, reagent grade, 97%
Supelco
Sodium formate, analytical standard
Supelco
Propionic acid, analytical standard
Sigma-Aldrich
Propionic acid, ≥99.5%
Sigma-Aldrich
Sodium formate, ACS reagent, ≥99.0%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%