Merck
Cellular origin and regulation of D- and L-serine in in vitro and in vivo models of cerebral ischemia.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (2014-10-09)
Takato Abe, Masataka Suzuki, Jumpei Sasabe, Shinichi Takahashi, Miyuki Unekawa, Kyoko Mashima, Takuya Iizumi, Kenji Hamase, Ryuichi Konno, Sadakazu Aiso, Norihiro Suzuki
ABSTRAKT
D-Serine is known to be essential for the activation of the N-methyl-D-aspartate (NMDA) receptor in the excitation of glutamatergic neurons, which have critical roles in long-term potentiation and memory formation. D-Serine is also thought to be involved in NMDA receptor-mediated neurotoxicity. The deletion of serine racemase (SRR), which synthesizes D-serine from L-serine, was recently reported to improve ischemic damage in mouse middle cerebral artery occlusion model. However, the cell type in which this phenomenon originates and the regulatory mechanism for D-/L-serine remain elusive. The D-/L-serine content in ischemic brain increased until 20 hours after recanalization and then leveled off gradually. The results of in vitro experiments using cultured cells suggested that D-serine is derived from neurons, while L-serine seems to be released from astroglia. Immunohistochemistry studies of brain tissue after cerebral ischemia showed that SRR is expressed in neurons, and 3-phosphoglycerate dehydrogenase (3-PGDH), which synthesizes L-serine from 3-phosphoglycerate, is located in astrocytes, supporting the results of the in vitro experiments. A western blot analysis showed that neither SRR nor 3-PGDH was upregulated after cerebral ischemia. Therefore, the increase in D-/L-serine was not related to an increase in SRR or 3-PGDH, but to an increase in the substrates of SRR and 3-PGDH.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
4-Fluoro-7-nitrobenzofurazan, ≥98% (elemental analysis)
Sigma-Aldrich
4-Fluoro-7-nitrobenzofurazan, suitable for fluorescence, BioReagent, ≥98.0% (HPLC)
Serine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DAPI, for nucleic acid staining
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Fluorescein, free acid