Merck
  • DPP IV inhibitor suppresses STZ-induced islets injury dependent on activation of the IGFR/Akt/mTOR signaling pathways by GLP-1 in monkeys.
DPP IV inhibitor suppresses STZ-induced islets injury dependent on activation of the IGFR/Akt/mTOR signaling pathways by GLP-1 in monkeys.
Biochemical and biophysical research communications (2014-12-03)
Yi Zhang, Younan Chen, Jingqiu Cheng, Zhiguang Guo, Yanrong Lu, Bole Tian
ABSTRAKT
To evaluate the protective effect of the DPP IV inhibitor in STZ-induced islet injury and to identify the molecular events that protect islet against apoptosis. 4 diabetic monkeys were treated with streptozotocin (70 mg/kg) in the presence or absence of the DPP IV inhibitor (Sitagliptin), continuing administered for 4 weeks after STZ. The monkeys were evaluated by plasma DPP IV activity, serum active GLP-1 response, blood glucose, insulin and C-P levels, the insulin resistance index (HOMA-IR), and the expression of insulin, caspase-3, IGF receptor (IGFR), p-Akt and p-mTOR in pancreas islets tissues. To test that DPP IV inhibitors might against islets apoptosis via IGFR/Akt/mTOR signaling pathways, the isolated islets from the normal monkeys were pre-treated with or without 10mM STZ for 1h, followed by GLP-1 (10 μM) in the presence or absence of NVP-AEW541 or Wortmannin for 24h, to determined islets function and islet apoptosis. DPP IV inhibitors treatment showed depressing the degradation of GLP-1 and significantly increased serum GLP-1 levels in DM monkeys. Moreover, treatment of diabetic monkeys with the DPP IV inhibitor or treatment of isolated islets with GLP-1 can decrease islet apoptosis, and enhanced islet function and survival, and the expression of IGF receptor, p-Akt and p-mTOR in islets. When the IGFR/Akt/mTOR signaling pathways was blocked by NVP-AEW541 or Wortmannin, the protective effects of GLP1 on STZ-induced islets injury were inhibited in vitro. Our data provides evidence that DPP IV inhibitors confer resistance to STZ-induced islet injury. The protective effects of DPP IV inhibitor on STZ-induced islets injury were dependent on activation of the IGFR/Akt/mTOR signaling pathways by GLP-1 in islets of monkeys.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Hexokinase from Saccharomyces cerevisiae, Type F-300, lyophilized powder, ≥130 units/mg protein (biuret)
Sigma-Aldrich
Hexokinase from Saccharomyces cerevisiae, lyophilized powder, ≥350 units/mg protein, Protein ≥10 % by biuret
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
Dithizone, Practical Grade
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dithizone, ACS reagent, for spectrophotometric det. of Cd, Cu, Hg, Pb, Zn, ≥98.0% (TLC)
USP
Dekstroza, United States Pharmacopeia (USP) Reference Standard