Merck
  • Frequencies of myohistological mitochondrial changes in patients with mitochondrial DNA deletions and the common m.3243A>G point mutation.
Frequencies of myohistological mitochondrial changes in patients with mitochondrial DNA deletions and the common m.3243A>G point mutation.
Neuropathology : official journal of the Japanese Society of Neuropathology (2014-11-08)
Charlotte Maria Zierz, Pushpa Raj Joshi, Stephan Zierz
ABSTRAKT
Frequencies of typical myohistological changes such as ragged red fibers (RRF) and cytochrome c oxidase (COX)-deficient fibers have been suggested to be dependent on underlying mitochondrial DNA (mtDNA) defect. However, there are no systematic studies comparing frequencies of myohistological changes and underlying genotypes. The histopathological changes were analysed in 29 patients with genetically confirmed mitochondrial myopathies. Genotypes included multiple mtDNA deletions due to POLG1 mutations (n = 11), single mtDNA deletion (n = 10) and mtDNA point mutation m.3243A>G (n = 8). Histochemical reactions, including Gomori-trichome, COX/SDH (succinate dehydrogenase) and SDH as well as immunohistological reaction with COX-antibody against subunit I (COI) were carried out in muscle biopsy sections of all patients. The COX-deficient fibers were observed most frequently in all three patient groups. The frequencies of myopathological changes were not significantly different in the different genotypes in all three histochemical stains. However, there was a tendency to lower means and variations in patients with point mutation. Only COI-negative fibers were histochemically negative for COX activity in all patient groups. Frequency of COI-negative fibers was significantly lower in patients with mtDNA point mutation than in patients with deletions. This suggests that impact of point mutation on protein synthesis is less than that of deletions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
USP
Alkohol odwodniony, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Disodium succinate, ≥98%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
USP
Kwas octowy lodowaty, United States Pharmacopeia (USP) Reference Standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Sodium succinate dibasic, purum, anhydrous, ≥98.0% (NT)