Merck
Molecularly imprinted solid-phase extraction for selective extraction of bisphenol analogues in beverages and canned food.
Journal of agricultural and food chemistry (2014-10-29)
Yunjia Yang, Jianlong Yu, Jie Yin, Bing Shao, Jing Zhang
ABSTRAKT
This study aimed to develop a selective analytical method for the simultaneous determination of seven bisphenol analogues in beverage and canned food samples by using a new molecularly imprinted polymer (MIP) as a sorbent for solid-phase extraction (SPE). Liquid chromatography coupled to triple-quadruple tandem mass spectrometry (LC-MS/MS) was used to identify and quantify the target analytes. The MIP-SPE method exhibited a higher level of selectivity and purification than the traditional SPE method. The developed procedures were further validated in terms of accuracy, precision, and sensitivity. The obtained recoveries varied from 50% to 103% at three fortification levels and yielded a relative standard deviation (RSD, %) of less than 15% for all of the analytes. The limits of quantification (LOQ) for the seven analytes varied from 0.002 to 0.15 ng/mL for beverage samples and from 0.03 to 1.5 ng/g for canned food samples. This method was used to analyze real samples that were collected from a supermarket in Beijing. Overall, the results revealed that bisphenol A and bisphenol F were the most frequently detected bisphenols in the beverage and canned food samples and that their concentrations were closely associated with the type of packaging material. This study provides an alternative method of traditional SPE extraction for screening bisphenol analogues in food matrices.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, analytical standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetrachlorobisphenol A, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
3,3′,5,5′-Tetrabromobisphenol A, 97%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
USP
Alkohol metylowy, United States Pharmacopeia (USP) Reference Standard
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis