Merck
  • Lack of interleukin-17 leads to a modulated micro-environment and amelioration of mechanical hypersensitivity after peripheral nerve injury in mice.
Lack of interleukin-17 leads to a modulated micro-environment and amelioration of mechanical hypersensitivity after peripheral nerve injury in mice.
Pain (2014-04-12)
Yuan-Ji Day, Jiin-Tarng Liou, Chiou-Mei Lee, Yi-Chiao Lin, Chih-Chieh Mao, An-Hsun Chou, Chia-Chih Liao, Hung-Chen Lee
ABSTRAKT
Interleukin-17 (IL-17) is involved in a wide range of inflammatory disorders and in recruitment of inflammatory cells to injury sites. A recent study of IL-17 knock-out mice revealed that IL-17 contributes to neuroinflammation and neuropathic pain after peripheral nerve injury. Surprisingly, little is known of micro-environment modulation by IL-17 in injured sites and in pathologically related neuroinflammation and chronic neuropathic pain. Therefore, we investigated nociceptive sensitization, immune cell infiltration, myeloperoxidase (MPO) activity, and expression of multiple cytokines and opioid peptides in damaged nerves of wild-type (IL-17(+/+)) and IL-17 knock-out (IL-17(-/-)) mice after partial sciatic nerve ligation. Our results demonstrated that the IL-17(-/-) mice had less behavioral hypersensitivity after partial sciatic nerve ligation, and inflammatory cell infiltration and pro-inflammatory cytokine (tumor necrosis factor-α, IL-6, and interferon-γ) levels in damaged nerves were significantly decreased, with the levels of anti-inflammatory cytokines IL-10 and IL-13, and expressions of enkephalin, β-endorphin, and dynorphin were also decreased compared to those in wild-type control mice. In conclusion, we provided evidence that IL-17 modulates the micro-environment at the level of the peripheral injured nerve site and regulates progression of behavioral hypersensitivity in a murine chronic neuropathic pain model. The attenuated behavioral hypersensitivity in IL-17(-/-) mice could be a result of decreased inflammatory cell infiltration to the injured site, resulting in modulation of the pro- and anti-inflammatory cytokine milieu within the injured nerve. Therefore, IL-17 may be a critical component for neuropathic pain pathogenesis and a novel target for therapeutic intervention for this and other chronic pain states.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Mouse Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Bovine TNFα / Tumor Necrosis Factor alpha ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Rat Tumor Necrosis Factor α ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant