Merck
  • Enantiomeric separation of tapentadol by capillary electrophoresis--study of chiral selectivity manipulation by various types of cyclodextrins.
Enantiomeric separation of tapentadol by capillary electrophoresis--study of chiral selectivity manipulation by various types of cyclodextrins.
Journal of pharmaceutical and biomedical analysis (2014-12-22)
Joanna Znaleziona, Ida Fejős, Juraj Ševčík, Michal Douša, Szabolcs Béni, Vítězslav Maier
ABSTRAKT
The chiral recognition of the centrally acting analgesic agent tapentadol and its isomers with various cyclodextrins (CDs) was studied by capillary electrophoresis, focusing on the migration order of four stereoisomers. In the case of non-charged hydroxypropylated CDs (2-hydroxypropyl-β-CD, 2-hydroxypropyl-γ-CD) the beta derivative was able to discriminate the S,R- and R,S-isomers in acidic background electrolyte, whereas the gamma allowed the separation of S,S- and R,R-tapentadol, respectively. Dual CD system containing both hosts was used to separate all of four isomers. Negatively charged sulfated-α-CD at 1.0% (w/v) concentration in 100mM sodium borate buffer (pH 9.5) was capable of separating the isomers with favorable enantiomer migration order and the optimized method was able to determine 0.15% of chiral impurities of tapentadol in the presence of the last migrating clinically important R,R-isomer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Lithium hydroxide, powder, reagent grade, ≥98%
Sigma-Aldrich
Lithium hydroxide, reagent grade, 98%
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)