Merck
Tuning of poly-S-nitrosated human serum albumin as superior antitumor nanomedicine.
Journal of pharmaceutical sciences (2014-05-23)
Yu Ishima, Jun Fang, Ulrich Kragh-Hansen, Hongzhuan Yin, Long Liao, Naohisa Katayama, Hiroshi Watanabe, Toshiya Kai, Ayaka Suenaga, Hiroshi Maeda, Masaki Otagiri, Toru Maruyama
ABSTRAKT
Macromolecules have been developed as carriers of low-molecular-weight drugs in drug delivery systems (DDS) to improve their pharmacokinetic profile or to promote their uptake in tumor tissue via enhanced permeability and retention (EPR) effects. We have previously demonstrated that poly-nitric oxide (NO) conjugated human serum albumin (Poly-SNO-HSA) has the potential to be a DDS carrier capable of accumulating NO in tumors. However, the stability of Poly-SNO-HSA in the circulation has to be improved, and its optimal molecular size for using the EPR effects has to be evaluated. In the present study, we performed two tuning methods for refining Poly-SNO-HSA, namely, pegylation and dimerization. We observed that pegylation enhanced the stability of Poly-SNO-HSA both in vitro and in vivo, and that dimerization of Poly-SNO-HSA enhanced the antitumor activity via more efficient delivery of NO in Colon 26 tumor-bearing mice. Intriguingly, dimerization resulted in a 10 times higher antitumor activity. These data suggest that pegylation and dimerization of Poly-SNO-HSA are very important tuners to optimize NO stability and accumulation, and thereby effect, in tumors. Thus, polyethylene glycol-Poly-SNO-HSA dimer seems to be a very appealing and safe NO carrier and thereby a strong candidate as an antitumor drug in future development of cancer therapeutics.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Isopentyl nitrite, 96%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
DL-Dithiothreitol, Molecular Biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
DL-Dithiothreitol, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥98% (titration)
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99% (titration)
Sigma-Aldrich
DL-Dithiothreitol, suitable for electrophoresis, ≥99%
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, suitable for suitability for complexometry, ≥99.0%
Sigma-Aldrich
DL-Dithiothreitol, BioUltra, Molecular Biology, ≥99.5% (RT)