Merck
  • UHPLC-MS/MS analysis of arachidonic acid and 10 of its major cytochrome P450 metabolites as free acids in rat livers: effects of hepatic ischemia.
UHPLC-MS/MS analysis of arachidonic acid and 10 of its major cytochrome P450 metabolites as free acids in rat livers: effects of hepatic ischemia.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2013-10-19)
Vindhya Edpuganti, Reza Mehvar
ABSTRAKT
The cytochrome P450 metabolites of arachidonic acid (AA) are mostly present in tissues, such as the liver, as bound to phospholipids, with only a small fraction available as free acids. The purpose of this study was to develop and validate a UHPLC-MS/MS method for quantitation of free liver concentrations of AA and four epoxygenated (5,6-, 8,9-, 11,12-, and 14,15-EET), four dihydroxylated (5,6-, 8,9-, 11,12-, and 14,15-DHET), and two ω/(ω-1) hydroxylated (19- and 20-HETEs) metabolites of AA in rat livers using deuterated internal standards. The analytes were rapidly and efficiently (79-92%) recovered from 100mg of fresh liver into methanol. After evaporation, the reconstituted samples were injected either undiluted (for the simultaneous analysis of the metabolites) into a gradient or diluted (for AA analysis) into an isocratic UHPLC system with run times of 5 and 2 min, respectively. Mass spectrometry was conducted using multiple reaction monitoring in negative mode. The method was linear (r(2)≥ 0.98) in the concentration ranges tested for metabolites (0.19-120 ng/g liver) and AA (7.8-500 μg/g liver). The lower limit of quantitation of the assay was between 0.57 and 5.6 pg injected on column for different AA metabolites. The assay was validated (n=5) based on acceptable intra- and inter-run accuracy and precision values. Additionally, matrix effect was minimal for most analytes. Freeze-thaw of samples drastically increased the free liver concentrations of analytes, presumably due to their release from the membrane storage sites. Therefore, fresh liver samples should be used for analysis. However, the methanolic extracts may be stored at -80°C for at least two weeks without any compromise. The method was successfully used in the measurement of all the analytes in the rats subjected to 60 min of hepatic ischemia (n=6) or sham operation (n=6). Ischemia resulted in significantly higher free concentrations of AA and most of its studied metabolites. The method is precise, accurate, and sensitive for measurement of free liver concentrations of AA and its P450 metabolites in the rat liver.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
20-Hydroxy-(5Z,8Z,11Z,14Z)-eicosatetraenoic acid, ~100 μg/mL in ethanol, ≥90% (HPLC)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
11,12-Epoxy-(5Z,8Z,14Z)-eicosatrienoic acid, ~100 μg/mL in ethanol, ≥95%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
USP
Kwas octowy lodowaty, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%