Merck
Sperm FISH and chromatin integrity in spermatozoa from a t(6;10;11) carrier.
Reproduction (Cambridge, England) (2014-04-10)
Marta Olszewska, Nataliya Huleyuk, Monika Fraczek, Danuta Zastavna, Ewa Wiland, Maciej Kurpisz
ABSTRAKT
Complex chromosome rearrangements (CCRs) are structurally balanced or unbalanced aberrations involving more than two breakpoints on two or more chromosomes. CCRs can be a potential reason for genomic imbalance in gametes, which leads to a drastic reduction in fertility. In this study, the meiotic segregation pattern, aneuploidy of seven chromosomes uninvolved in the CCR and chromatin integrity were analysed in the ejaculated spermatozoa of a 46,XY,t(6;10;11)(q25.1;q24.3;q23.1)mat carrier with asthenozoospermia and a lack of conception. The frequency of genetically unbalanced spermatozoa was 78.8% with a prevalence of 4:2 segregants of 38.2%, while the prevalence of the adjacent 3:3 mode was 35.3%. Analysis of the aneuploidy of chromosomes 13, 15, 18, 21, 22, X and Y revealed an approximately fivefold increased level in comparison with that of the control group, indicating the presence of an interchromosomal effect. Sperm chromatin integrity status was evaluated using chromomycin A3 and aniline blue staining (deprotamination), acridine orange test and TUNEL assay (sperm DNA fragmentation). No differences were found when comparisons were made with a control group. We suggest that the accumulation of genetically unbalanced spermatozoa, significantly increased sperm aneuploidy level and decreased sperm motility (20%, progressive) were not responsible for the observed lack of reproductive success in the analysed infertile t(6;10;11) carrier. Interestingly, in the case described herein, a high level of sperm chromosomal imbalance appears not to be linked to sperm chromatin integrity status.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)