Merck
Click conjugation of peptide to hydrogel nanoparticles for tumor-targeted drug delivery.
Biomacromolecules (2014-08-28)
Ming Qin, Hong Zong, Raoul Kopelman
ABSTRAKT
Here we introduce a modified peptide-decorated polymeric nanoparticle (NP) for cancer cell targeting, which can deliver drugs, such as doxorubicin (Dox), to several kinds of cancer cells. Specifically, we employ a nucleolin-targeting NP, with a matrix based on a copolymer of acrylamide (AAm) and 2-carboxyethyl acrylate (CEA). The negatively charged co(CEA-AAm) NP was conjugated with a nucleolin-targeting F3 peptide using a highly efficient and specific copper(I) catalyzed azide-alkyne click reaction. F3 peptide binds to angiogenic tumor vasculatures and other nucleolin overexpressing tumor cells. Attaching F3 peptide onto the NP increases the NP uptake by the nucleolin-expressing glioma cell line 9L and the breast cancer cell line MCF-7. Notably, the F3-conjugated NPs show much higher uptake by the nucleolin-overexpressing glioma cell line 9L than that by the breast cancer cell line MCF-7, the latter having a lower expression of nucleolin on its plasma membrane surface. Moreover, the F3 peptide also dramatically enhances the uptake of co(CEA-AAm) NPs by the drug-resistant cell line NCI/ADR-RES. Also, with this F3-conjugated co(CEA-AAm) NP, a high loading and slow release of doxorubicin were achieved.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Alkohol odwodniony, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Carboxyethyl acrylate, contains 900-1100 ppm MEHQ as inhibitor
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Copper(II) sulfate, ReagentPlus®, ≥99%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, BioUltra, ≥98.0% (RT)