Merck
Certification of a reference material of metal content in atmospheric particles deposited on filters.
Analytical and bioanalytical chemistry (2014-09-28)
Caroline Oster, Guillaume Labarraque, Paola Fisicaro
ABSTRAKT
Air quality is one of the areas in Europe where a series of EU Directives have been published with the aim of achieving improved long-term and harmonised air quality objectives across the European Union. This paper describes the production of a certified reference material, aiming to support QA/QC programmes of analytical laboratories in the framework of the air quality monitoring activities. The certified values are the As, Cd, Ni and Pb masses in PM10 particles deposited on quartz filters (CRM SL-MR-2-PSF-01). All the steps of the certification, i.e. the material characterisation, homogeneity and stability evaluation and uncertainty calculation, were performed according to the ISO guide 35 guidelines. The certification was conducted using the characterisation by a single method approach based on isotope dilution for cadmium, nickel, and lead and gravimetric standard addition calibration for arsenic associated with inductively coupled mass spectrometry (ICP-MS). The amounts of the four elements are in the range of the target values regulated by EU Directives.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Boric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Boric acid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Boric acid, BioReagent, Molecular Biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Supelco
Benalaxyl-M, PESTANAL®, analytical standard
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Supelco
Koncentrat kwasu azotowego, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Boric acid, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Boric acid, ACS reagent, ≥99.5%