Merck
Effect of mushroom diet on pharmacokinetics of gabapentin in healthy Chinese subjects.
British journal of clinical pharmacology (2013-10-31)
Dorothy Su Lin Toh, Lie Michael George Limenta, Jie Yin Yee, Ling-Zhi Wang, Boon-Cher Goh, Michael Murray, Edmund Jon Deoon Lee
ABSTRAKT
This study evaluated the pharmacokinetics of gabapentin in Chinese subjects who received a diet rich in shiitake mushrooms. Shiitake mushrooms have been shown to contain high amount of ergothioneine. In vitro studies have shown that OCTN1-mediated secretion of gabapentin is trans-stimulated by ergothioneine. This study also investigated the concentrations of ergothioneine in plasma at baseline and following mushroom consumption. Ten healthy male subjects were recruited and received a diet containing no mushrooms (treatment A) or a high mushroom diet (treatment B; after at least a 7 day washout period) 1 day prior to administration of a single oral dose of gabapentin 600 mg. Ingestion of shiitake mushrooms produced significant increases in plasma ergothioneine concentrations that were sustained for more than 48 h. A statistically significant but modest increase in the renal clearance (CLR ) of gabapentin occurred after intake of the mushroom diet (91.1 ± 25.1 vs. 76.9 ± 20.6 ml min(-1) , P = 0.031). No significant changes in AUC(0,tlast ) of gabapentin were observed (P = 0.726). Creatinine clearance did not correlate with CLR of gabapentin at baseline (treatment A). After ingestion of the mushroom diet, creatinine clearance accounted for 65.3% of the variance in CLR of gabapentin. These data suggest that diet-drug pharmacokinetic interactions may occur during co-exposure to gabapentin and mushroom constituents. However, as it does not affect the AUC(0,tlast ) of gabapentin, it may not have clinically important consequences. Shiitake mushrooms can also be used as a source of ergothioneine for future clinical studies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
L-(+)-Ergothioneine
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%