Merck
Mesoporous silica coated gold nanorods loaded doxorubicin for combined chemo-photothermal therapy.
International journal of pharmaceutics (2014-05-06)
A Soltan Monem, Nihal Elbialy, Noha Mohamed
ABSTRAKT
The efficacy of the combined chemo-photothermal therapy, using a mesoporous silica-coated gold nanorods loaded DOX (pGNRs@mSiO2-DOX), was consistently tested both in vitro and in vivo. The prepared nanoparticles that were characterized using transmission electron microscopy (TEM), UV-vis absorption spectroscopy and zeta potential showed high doxorubicin loading capacity in addition to its pH-responsive release. The pGNRs@mSiO2-DOX photo-heat conversion characteristic found to be stable for several repeated NIR irradiated doses was tested in simulated body fluid. In vitro results showed that pGNRs@mSiO2-DOX causes a significant damage in breast cancer cell line MCF-7 compared to free DOX. Contrary to this, it showed low toxicity to human amnion wish cells compared to CTAB coated GNRs and free DOX. In vivo results showed that intravenous administration of pGNRs@mSiO2-DOX (1.7 mg/kg) markedly suppresses the growth of subcutaneous Ehrlich carcinoma in female Balb mice (p<0.0001). Consistently, histopathological examination revealed a complete loss of tumor cellular details for mice that received the combined treatment. Based on the obtained results, this passively targeted pGNRs@mSiO2-DOX could specifically deliver drug and excessive local heat to tumor sites achieving high combined therapeutic efficacy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ammonium bromide, 99.999% trace metals basis
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis