Merck
Glycogen synthase kinase 3 regulates IL-1β mediated iNOS expression in hepatocytes by down-regulating c-Jun.
Journal of cellular biochemistry (2014-08-28)
Jaganathan Lakshmanan, Baochun Zhang, Ikenna C Nweze, Yibo Du, Brian G Harbrecht
ABSTRAKT
Excessive nitric oxide from the inducible nitric oxide synthase (iNOS) increases shock-induced hepatic injury, hepatic dysfunction, inflammation, and mortality in animal models. Cytokines increase the expression of iNOS in hepatocytes, but the signaling mechanisms involved are not completely understood. We have previously demonstrated that Akt mediates the inhibitory effect of cAMP and insulin on cytokine-induced hepatocyte iNOS expression. We hypothesized that glycogen synthase kinase 3 (GSK3), a target of Akt phosphorylation, would regulate hepatocyte iNOS expression. In cultured rat hepatocytes, GSK3 inhibitors decreased IL-1β mediated nitric oxide (NO) production and iNOS protein expression, while the phosphatidylinositol 3-kinase (PI3K)/Akt pathway inhibitor LY294002 increased the cytokine-mediated NO production and iNOS expression. Over-expression of the constitutively active form of GSK3β enhanced IL-1β-mediated iNOS expression. GSK3 catalyzes the phosphorylation of c-Jun at the c-terminal Thr239 that facilitates c-Jun degradation. Inhibition of GSK3 with SB216763 and lithium chloride significantly reduced, whereas blocking PI3K/Akt increased phosphorylation of c-Jun at Thr239. The levels of total-c-Jun and c-Jun phosphorylated at Ser63 inversely correlated with c-Jun phosphorylated at Thr239, GSK3 activation and iNOS expression. Over-expression of a dominant negative c-Jun not only caused an increase in IL-1β-mediated iNOS promoter activity and iNOS protein expression but was also able to reverse the SB216763-mediated suppression of iNOS. These results demonstrate that GSK3, a downstream target of Akt, regulates IL-1β-stimulated iNOS expression in hepatocytes by directly phosphorylating c-Jun in an inhibitory manner.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
NGF-β human, from human, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture
Sigma-Aldrich
TDZD-8, ≥98% (HPLC), needles
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Supelco
Lithium chloride solution, 1 M in ethanol
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Lithium chloride solution, 2 M in ethanol
SAFC
L-Glutamine
Supelco
Electrolyte solution, nonaqueous, 2 M LiCl in ethanol
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)