Merck
Omega-3 fatty acids inhibit the up-regulation of endothelial chemokines in maintenance hemodialysis patients.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association (2014-09-11)
Adriana M Hung, Cindy Booker, Charles D Ellis, Edward D Siew, Amy J Graves, Ayumi Shintani, Naji N Abumrad, Jonathan Himmelfarb, Talat Alp Ikizler
ABSTRAKT
Chronic systemic inflammation is common in patients with chronic kidney disease on dialysis (CKD5D) and has been considered a key mediator of the increased cardiovascular risk in this patient population. In this study, we tested the hypothesis that supplementation of omega-3 polyunsaturated fatty acids (ω-3 PUFAs) will attenuate the systemic inflammatory process in CKD5D patients. The design was a randomized, double-blinded, placebo controlled pilot trial (NCT00655525). Thirty-eight patients were randomly assigned in a 1 : 1 fashion to receive 2.9 g of eicosapentaenoic acid (C20:5, n-3) plus docosahexaenoic acid (C22:6, n-3) versus placebo for 12 weeks. The primary outcome was change in pro-inflammatory chemokines measured by lipopolysaccharide (LPS)-stimulated peripheral blood mononuclear cells (PBMCs). Secondary outcomes were changes in systemic inflammatory markers. Analysis of covariance was used to compare percent change from baseline to 12 weeks. Thirty-one patients completed 12 weeks and three patients completed 6 weeks of the study. Median age was 52 (interquartile range 45, 60) years, 74% were African-American and 79% were male. Supplementation of ω-3 PUFAs effectively decreased the LPS-induced PBMC expression of RANTES (Regulated upon Activation, Normal T cell Expressed and Secreted) and MCP-1 (Monocyte Chemotactic Protein-1; unadjusted P = 0.04 and 0.06; adjusted for demographics P = 0.02 and 0.05, respectively). There was no significant effect of the intervention on serum inflammatory markers (C-reactive protein, interleukin-6 and procalcitonin). The results of this pilot study suggest that supplementation of ω-3 PUFAs is beneficial in decreasing the levels of endothelial chemokines, RANTES and MCP-1. Studies of larger sample size and longer duration are required to further evaluate effects of ω-3 PUFAs on systemic markers of inflammation, other metabolic parameters and clinical outcomes, particularly cardiovascular outcomes in CKD5D patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
(L)-Dehydroascorbic acid
Sigma-Aldrich
Dehydro-L-(+)-ascorbic acid dimer, ≥80% (enzymatic)
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dekstroza, United States Pharmacopeia (USP) Reference Standard