Merck
Tumour-associated fibroblasts contribute to neoangiogenesis in human parathyroid neoplasia.
Endocrine-related cancer (2014-12-18)
C Verdelli, L Avagliano, P Creo, V Guarnieri, A Scillitani, L Vicentini, G B Steffano, E Beretta, L Soldati, E Costa, A Spada, G P Bulfamante, S Corbetta
ABSTRAKT
Components of the tumour microenvironment initiate and promote cancer development. In this study, we investigated the stromal component of parathyroid neoplasia. Immunohistochemistry for alpha-smooth muscle actin (α-SMA) showed an abundant periacinar distribution of α-SMA(+) cells in normal parathyroid glands (n=3). This pattern was progressively lost in parathyroid adenomas (PAds; n=6) where α-SMA(+)cells were found to surround new microvessels, as observed in foetal parathyroid glands (n=2). Moreover, in atypical adenomas (n=5) and carcinomas (n=4), α-SMA(+) cells disappeared from the parenchyma and accumulated in the capsula and fibrous bands. At variance with normal glands, parathyroid tumours (n=37) expressed high levels of fibroblast-activation protein (FAP) transcripts, a marker of tumour-associated fibroblasts. We analysed the ability of PAd-derived cells to activate fibroblasts using human bone-marrow mesenchymal stem cells (hBM-MSCs). PAd-derived cells induced a significant increase in FAP and vascular endothelial growth factor A (VEGFA) mRNA levels in co-cultured hBM-MSCs. Furthermore, the role of the calcium-sensing receptor (CASR) and of the CXCL12/CXCR4 pathway in the PAd-induced activation of hBM-MSCs was investigated. Treatment of co-cultures of hBM-MSCs and PAd-derived cells with the CXCR4 inhibitor AMD3100 reduced the stimulated VEGFA levels, while CASR activation by the R568 agonist was ineffective. PAd-derived cells co-expressing parathyroid hormone (PTH)/CXCR4 and PTH/CXCL12 were identified by FACS, suggesting a paracrine/autocrine signalling. Finally, CXCR4 blockade by AMD3100 reduced PTH gene expression levels in PAd-derived cells. In conclusion, i) PAd-derived cells activated cells of mesenchymal origin; ii) PAd-associated fibroblasts were involved in tumuor neoangiogenesis and iii) CXCL12/CXCR4 pathway was expressed and active in PAd cells, likely contributing to parathyroid tumour neoangiogenesis and PTH synthesis modulation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
AMD3100 octahydrochloride hydrate, ≥97% (NMR), solid
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - FITC antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland