Merck
  • Fabric phase sorptive extraction: a new sorptive microextraction technique for the determination of non-steroidal anti-inflammatory drugs from environmental water samples.
Fabric phase sorptive extraction: a new sorptive microextraction technique for the determination of non-steroidal anti-inflammatory drugs from environmental water samples.
Analytica chimica acta (2015-03-04)
Inés Racamonde, Rosario Rodil, José Benito Quintana, Benigno José Sieira, Abuzar Kabir, Kenneth G Furton, Rafael Cela
ABSTRAKT
Fabric phase sorptive extraction (FPSE) is a new, yet very promising member of the sorbent-based sorptive microextraction family. It has simultaneously improved both the extraction sensitivity and the speed of the extraction by incorporating high volume of sol-gel hybrid inorganic-organic sorbents into permeable fabric substrates. The advantages of FPSE have been investigated for the determination of four non-steroidal anti-inflammatory drugs, ibuprofen, naproxen, ketoprofen and diclofenac, in environmental water samples in combination with gas chromatography-mass spectrometry. Initially, the significance of several parameters affecting FPSE: sorbent chemistry, matrix pH and ionic strength were investigated using a mixed level factorial design (3(1)×2(2)). Then, other important parameters e.g., sample volume, extraction kinetics, desorption time and volume were also carefully studied and optimized. Due to the high sorbent loading on the FPSE substrate in the form of ultra-thin coating and the open geometry of the microextraction device, higher mass transfer of the target analytes occurs at a faster rate, leading to high enrichment factors in a relatively short period of time (equilibrium times: 45-100 min). Under optimal operational conditions, the limits of detection (S/N=3) were found to be in the range of 0.8 ng L(-1) to 5 ng L(-1). The enrichment factors ranged from 162 to 418 with absolute extraction efficiencies varied from 27 to 70%, and a good trueness (82-116% relative recoveries) indicating that the proposed method can be readily deployed to routine environmental pollution monitoring. The proposed method was successfully applied to the analysis of target analytes in two influent and effluent samples from a wastewater treatment plant and two river water samples in Spain.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
N-Acetyl-D-penicillamine, ≥99.0% (T)
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Informacje o cenach i dostępności nie są obecnie dostępne.
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, LiChropur, ≥95.0% (GC)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, ≥95%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, pink hub
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
SPME Portable Field Sampler, coating PDMS
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, yellow hub
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, yellow hub
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, green hub
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, green hub
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, white hub
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Informacje o cenach i dostępności nie są obecnie dostępne.
SAFC
Sodium chloride solution, 5 M
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
SPME Fiber Assembly, 100 μm PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, red hub
Informacje o cenach i dostępności nie są obecnie dostępne.