Merck
In vitro anti-aging activities of extracts from leaves of Ma Kiang (Cleistocalyx nervosum var. paniala).
Pharmaceutical biology (2014-11-29)
Jiradej Manosroi, Charinya Chankhampan, Kulthida Kumguan, Worapaka Manosroi, Aranya Manosroi
ABSTRAKT
Cleistocalyx nervosum (DC.) Kosterm. var. paniala (Roxb.) J. Parn. & Chantaran. (Myrtaceae) or Ma Kiang contains high amounts of phenolic compounds. Antioxidant activity of its fruit and seed has been investigated. However, limited available information concerning the biological activities of its leaves has been reported. To investigate the in vitro anti-aging potential of young and old leaves of Ma Kiang. Ma Kiang leaves were extracted using water, methanol, and chloroform as the solvents by cold (sonication) and hot (boiling) processes. The extracts were determined for total phenolic and flavonoid contents. The extracts (at 0.001-10 mg/ml) were tested for antioxidative and tyrosinase inhibition activities using a colorimetric method. The cytotoxicity of extracts (at 0.0001-1 mg/ml) was determined with human skin fibroblasts. Also, the extracts at 0.001, 0.01, and 0.1 mg/ml which showed no toxicity were tested for MMP-2 inhibition. The cold methanol extract of the old leaves showed the highest total phenolic and flavonoid contents of 511.44 ± 18.23 μg GAE/mg and 262.96 ± 2.98 μg QE/mg, respectively. This extract also gave high free radical scavenging, lipid peroxidation inhibition, and tyrosinase inhibition activities with SC50, IPC50, and IC50 values of 0.02 ± 0.004, 0.23 ± 0.13, and 0.02 ± 0.006 mg/ml, respectively. The extract at 0.1 mg/ml exhibited the highest MMP-2 inhibition of 91.14 ± 1.67%. The anti-aging potential of the cold methanol extract from old leaves of Ma Kiang can be further developed as an anti-aging agent.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
USP
Alkohol odwodniony, United States Pharmacopeia (USP) Reference Standard
Supelco
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(±)-α-Tocopherol, analytical standard
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
(±)-α-Tocopherol, tested according to Ph. Eur.
SAFC
L-Tyrosine
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Aluminum chloride, ReagentPlus®, 99%
Sigma-Aldrich
Aluminum chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, reagent grade, 98%
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride solution, 1.0 M in nitrobenzene
USP
L-Tyrosine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Ascorbic acid, reagent grade