Merck
Autologous platelet gel to treat chronic decubital ulcers: a randomized, blind controlled clinical trial in dogs.
Veterinary surgery : VS (2014-02-04)
Adolfo Maria Tambella, Anna Rita Attili, Fabrizio Dini, Angela Palumbo Piccionello, Cecilia Vullo, Evelina Serri, Paolo Scrollavezza, Gilles Dupré
ABSTRAKT
To determine the efficacy of topical application of the autologous platelet gel (PG) in canine chronic protracted decubital ulcers. Prospective, randomized, blind controlled clinical trial. Dogs (n = 18) with bilateral chronic wounds caused by protracted decubitus ulcers. For each dog, wound side was randomized to receive either platelet gel (group PG) every 5 days for 5 dressing changes, or paraffin gauzes dressings (group C), as negative control. Wound healing and wound surfaces were compared at admission and then evaluated every 5th day, until day 25. Outcome variables were: open wound area, reduction of open wound surface compared to admission and to each preceding dressing change, time to complete epithelialization. Significant differences in healing process were observed at day 5 and continued throughout the entire study period (P < .00001). At 25 days, mean percent reduction in wound area was 93.5% in group PG and 13.2% in group C (P < .00001). Appropriately prepared autologous PG, an inexpensive, readily available blood derivative, applied topically results in more rapid healing of chronic non-healing decubital ulcers in dogs than those treated by use of paraffin-impregnated gauzes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Mannitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dekstroza, United States Pharmacopeia (USP) Reference Standard