Merck
Ultrasensitive SERS detection of VEGF based on a self-assembled Ag ornamented-AU pyramid superstructure.
Biosensors & bioelectronics (2015-02-04)
Sen Zhao, Wei Ma, Liguang Xu, Xiaoling Wu, Hua Kuang, Libing Wang, Chuanlai Xu
ABSTRAKT
For the first time, we demonstrated the fabrication of silver nanoparticle ornamented-gold nanoparticle pyramids (Ag-Au Pys) using an aptamer-based self-assembly process and investigated their surface-enhanced Raman scattering (SERS) properties in the detection of vascular endothelial growth factor (VEGF). Under optimized conditions, the SERS signal was negatively related to VEGF concentration over the range 0.01-1.0 fM and the limit of detection (LOD) was as low as 22.6 aM. The matrix effect and the specificity of this developed method were further examined, and the results showed that the superstructure sensor was ultrasensitive and highly selective. This developed aptamer-based SERS detection method suggests that it may be a promising strategy for a variety of sensing applications.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, ≥99.999% trace metals basis
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
USP
Tromethamine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
4-Nitrothiophenol, technical grade, 80%
SAFC
Tromethamine
Trometamol, European Pharmacopoeia (EP) Reference Standard