Merck
  • Post-acquisition analysis of untargeted accurate mass quadrupole time-of-flight MS(E) data for multiple collision-induced neutral losses and fragment ions of glutathione conjugates.
Post-acquisition analysis of untargeted accurate mass quadrupole time-of-flight MS(E) data for multiple collision-induced neutral losses and fragment ions of glutathione conjugates.
Rapid communications in mass spectrometry : RCM (2014-11-08)
Andreas Brink, Fabien Fontaine, Michaela Marschmann, Bernd Steinhuber, Esra Nurten Cece, Ismael Zamora, Axel Pähler
ABSTRAKT
Analytical methods to assess glutathione (GSH) conjugate formation based on mass spectrometry usually take advantage of the specific fragmentation behavior of the glutathione moiety. However, most methods used for GSH adduct screening monitor only one specific neutral loss or one fragment ion, even though the peptide moiety of GSH adducts shows a number of other specific neutral fragments and fragment ions which can be used for identification. Nine reference drugs well known to form GSH adducts were incubated with human liver microsomes. Mass spectrometric analysis was performed with a quadrupole time-of-flight mass spectrometer in untargeted accurate mass MS(E) mode. The data analysis and evaluation was achieved in an automated approach with software to extract and identify GSH conjugates based on the presence of multiple collision-induced neutral losses and fragment ions specific for glutathione conjugates in the high-energy MS spectra. In total 42 GSH adducts were identified. Eight (18%) adducts did not show the neutral loss of 129 but were identified based on the appearance of other GSH-specific neutral losses or fragment ions. In high-energy MS(E) spectra the GSH-specific fragment ions of m/z 308 and 179 as well as the neutral loss of 275 Da were complementary to the commonly used neutral loss of 129 Da. Further, one abundant (yet unpublished) GSH conjugate of troglitazone formed in human liver microsomes was found. A software-aided approach was developed to reliably retrieve GSH adduct formation data out of untargeted complex full scan QTOFMS(E) data in a fast and efficient way. The present approach to detect and analyze multiple collision-induced neutral losses and fragment ions of glutathione conjugates in untargeted MS(E) data might be applicable to higher throughput to assess reactive metabolite formation in drug discovery.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
DL-Leucine, ≥99% (HPLC)
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Acetaminofen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Troglitazone, ≥98% (HPLC)
Supelco
Water, suitable for ion chromatography
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture