Merck
Mast cells, neovascularization, and microhemorrhages are associated with saccular intracranial artery aneurysm wall remodeling.
Journal of neuropathology and experimental neurology (2014-08-08)
Eliisa Ollikainen, Riikka Tulamo, Juhana Frösen, Satu Lehti, Petri Honkanen, Juha Hernesniemi, Mika Niemelä, Petri T Kovanen
ABSTRAKT
Chronic inflammation contributes to remodeling, degeneration, and rupture of saccular intracranial artery aneurysms. Mast cells are important proinflammatory and proangiogenic cells in chronic inflammatory vascular diseases. Here we studied mast cells and neovascularization in 36 intraoperatively resected aneurysms using histology and immunohistochemistry and analyzed the clinical characteristics of the aneurysms according to bleeding status (unruptured vs ruptured). Among the 36 aneurysms, 9 contained mast cells (tryptase-positive cells) and 15 contained neovessels (CD34- and CD31-positive capillarylike structures). The density of neovessels was significantly higher in aneurysm walls containing mast cells than in walls not containing them. In particular, wall areas with abundant mast cells and neovessels also contained iron deposits, indicating damage of newly formed endothelium with ensuing microhemorrhages. Walls with the highest neovessel density and the greatest iron deposition also showed evidence of degeneration. Finally, none of the mast cell-containing aneurysms showed an intact luminal endothelium. Thus, mast cells may adversely affect both neovascular and luminal endothelia. The novel association of mast cells with neovessels and injurious microhemorrhages, as well as with luminal endothelial erosion, suggests that mast cells contribute to remodeling and degeneration of saccular intracranial artery aneurysms.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glikol propylenowy, United States Pharmacopeia (USP) Reference Standard
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC