Merck
Cytoplasmic interaction of the tumour suppressor protein hSNF5 with dynamin-2 controls endocytosis.
Oncogene (2013-07-16)
T Alfonso-Pérez, M S Domínguez-Sánchez, M García-Domínguez, J C Reyes
ABSTRAKT
Human SNF5 (hSNF5; INI1, SMARCB1 or BAF47) is a component of the human SWI/SNF chromatin remodelling complex and a tumour suppressor mutated in rhabdoid tumours. It also associates with the integrase of the human immunodeficiency virus (HIV)-1. We show by fluorescence loss in photobleaching that hSNF5 is constantly shuttling between the nucleus and the cytoplasm, raising the question of what the role of hSNF5 is in the cytoplasm. Here, we demonstrate that hSNF5 directly interacts with the GTPase dynamin-2 (DNM2) in the cytoplasm. DNM2 is a large GTPase involved in endocytosis and vesicle dynamics, which has been related to HIV-1 internalization. We show that hSNF5 colocalizes with DNM2 in endocytic vesicles. Depletion of hSNF5, but not of other components of the SWI/SNF complex, destabilizes DNM2 and impairs DNM2-dependent endocytosis. Furthermore, we show that hSNF5 inhibits assembly-stimulated DNM2 GTPase activity but not basal GTPase activity in vitro. Altogether, these results indicate that hSNF5 affects both the stability and the activity of DNM2, uncovering an unexpected role of hSNF5 in modulating endocytosis, and open new perspectives in understanding the role of hSNF5 in tumour genesis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
PIPES, anhydrous, free-flowing, Redi-Dri, ≥99%
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
PIPES, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
PIPES, BioXtra, ≥99% (titration)
Sigma-Aldrich
PIPES, ≥99% (titration)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
PIPES, BioXtra, Molecular Biology, ≥99.5% (T)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)