Merck
Stability characterization and appearance of particulates in a lyophilized formulation of a model peptide hormone-human secretin.
International journal of pharmaceutics (2015-02-01)
Charudharshini Srinivasan, Akhtar Siddiqui, Maxwell Korang-Yeboah, Mansoor A Khan
ABSTRAKT
Drug shortages and recalls are often caused due to particulate growth in parenteral products and can have serious clinical implications. Root cause analysis of such recalls and shortages may arise due to insufficient understanding of process, formulations issues and environmental effects than often reported filtration and inadequate personnel training. Therefore, the goal of this study was to use a model peptide hormone, secretin that is currently under drug shortage, and investigate the effect of excipients on the lyophilized secretin formulation and evaluate the effect of storage and excursion temperatures. Lyophilized formulation was assayed for secretin by reverse phase HPLC. Solid state characteristics of lyophilized formulation were determined by X-ray powder diffraction (XRPD), thermal and spectroscopic methods. Dynamic light scattering (DLS) was used to detect particulates in the formulation after reconstitution. To assess the environmental impact, the lyophilized samples were stored at -20°C, 4°C, 25°C and 25°C/60%RH and analyzed at time 0, 1, 4, and 8 weeks. HPLC analyses exhibited a decrease in secretin concentration by 8 week (20-27% fold decrease). Visual observation and DLS showed particulates and increased reconstitution time (e.g., at 25°C/60%RH, particle size of ∼390 nm at day 0 to >2 μm as early as week 1; reconstitution time of ∼20s at day 0 to ∼67s at week 8). XRPD, thermal and spectroscopic methods demonstrated polymorphic transitions of mannitol and increased crystallinity in the lyophilized formulations with time. These studies potentially address the effect of product excursions outside the proposed label storage conditions which is -20°C for secretin formulation and this is the first time it has been investigated. These observations indicate that both environmental factor and excipient may have an impact on the stability of secretin formulation and appearance of particles in the product.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Mannitol, European Pharmacopoeia (EP) Reference Standard
SAFC
Sodium chloride solution, 5 M
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.