Merck
Regeneration of spermatogenesis in a mouse model of azoospermia by follicle-stimulating hormone and oestradiol.
Andrologia (2013-12-12)
A Jafarian, M R Sadeghi, N Pejhan, S Salehkhou, N Lakpour, M M Akhondi
ABSTRAKT
Busulfan is a chemotherapeutic drug that induces sterility, azoospermia and testicular atrophy. To induce degeneration of spermatogenesis, we used different amounts of busulfan. Adult male C57Bl/6 mice were treated with 15, 30 and 45 mg kg(-1) of busulfan. After 5 weeks, animals had daily injections of 7.5 IU human follicle-stimulating hormone (hFSH) and 12.5 μg kg(-1) oestradiol benzoate (EB), separately or simultaneously. After this time, the animals were killed and blood samples were taken through cardiac puncture. Testes were used for histopathology experiments, DNA flow cytometry and RNA extraction for expression of c-kit and cyclin B1 genes. EB unlike FSH has induced stimulatory effects on spermatogenesis, increased the level of serum testosterone 2-fold and caused a 2-fold increase in the number of haploid cells. The result showed that hFSH with EB multiplied EB stimulatory effects on spermatogenesis up to four times. Expression of c-kit and cyclin B1 genes increased in EB and hFSH+EB groups. These findings suggest that EB regulates spermatogonial stem cells via hFSH. hFSH with EB had synergistic effect on regeneration of spermatogenesis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Supelco
Busulfan, analytical standard, for drug analysis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis